missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Buffered Peptone Water Preenrichment

For injured Salmonella in food

Manufacturer:  BD B218104

 View more versions of this product

Catalog No. DF1810-08-0


 

Specifications Description & Specifications

Specifications

ISO/DIS, 11014, BAM, CCAM, ISO, USDA
Dehydrated
Peptone Water
Peptone Water, Buffered Pre-Enrichment
<30°C
Isolation of Salmonella species
Peptone 10g/L, Sodium Chloride 5g/L, Disodium Phosphate 3.5g/L, Monopotassium Phosphate 1.5g/L
Bottle
10kg