Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Brain Heart Infusion Agar


For isolation of Aspergillus brasiliensis (niger) 16404, Escherichia coli 25922, Staphylococcus aureus 25923, Streptococcus pneumoniae 6305, Streptococcus pyogenes 19615

Manufacturer: bd  B241820

 View more versions of this product

Catalog No. DF0418-15-9

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO, 9000
Format Dehydrated
For Use With (Application) Isolation of Aspergillus brasiliensis (niger) 16404, Escherichia coli 25922, Staphylococcus aureus 25923, Streptococcus pneumoniae 6305, Streptococcus pyogenes 19615
Storage Requirements <30°C
Product Type Brain heart infusion Agar
Packaging Bottle
Quantity 100g
Shelf Life 1825 Days
Media Format Calf brain infusion from 200g 7.7g/L, Beef heart infusion from 250g 9.8g/L, Proteose Peptone 10g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 15g/L
Media Type BHI Agar
Accessories

1825 Days