Please login to your online account to display your discounted pricing

BD™ Difco™ Dehydrated Culture Media: Brain Heart Infusion Agar


For isolation of Aspergillus brasiliensis (niger) 16404, Escherichia coli 25922, Staphylococcus aureus 25923, Streptococcus pneumoniae 6305, Streptococcus pyogenes 19615

Manufacturer: bd™  B241810

 View more versions of this product

Catalog No. DF0418-07-9

  • / Each
 

Description & Specifications

Specifications

Color Beige
Storage Requirements <30°C
Certifications/Compliance ISO 9000, AOAC, BAM, CCAM, COMPF, EPA, MCM9, SMD, SMWW, USDA
Format Dehydrated
Quantity 2kg
Media Type BHI Agar
Media Format Calf brain infusion from 200g 7.7g/L, Beef heart infusion from 250g 9.8g/L, Proteose Peptone 10g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 15g/L
Product Type Brain heart infusion Agar
Packaging Pail
For Use With (Application) Isolation of Aspergillus brasiliensis (niger) 16404, Escherichia coli 25922, Staphylococcus aureus 25923, Streptococcus pneumoniae 6305, Streptococcus pyogenes 19615
Shelf Life 1825 Days

BHI Agar has proven to be effective in the cultivation of a wide variety of fastidious microorganisms, including many types of pathogens, fungi and yeasts. Also used with added antibiotics in isolating fungi. It derives its nutrients from the brain heart infusion, peptone and dextrose components.

  • The peptones and infusion are sources of organic nitrogen, carbon, sulfur, vitamins and trace substances
  • Dextrose is a carbohydrate source that microorganisms utilize by fermentative action
  • The medium is buffered through the use of disodium phosphate
  • To prepare solution, suspend 52g in 1L purified water
  • pH (5.2% solution, 25°C): 7.4 ± 0.2
  • Good cultural response with: Aspergillus brasiliensis (niger) 16404, Escherichia coli 25922, Staphylococcus aureus 25923, Streptococcus pneumoniae 6305, Streptococcus pyogenes 19615

Accessories

1825 Days