Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Bile Salts No. 3


Manufacturer: bd  B213010

 View more versions of this product

Catalog No. DF0130-15-6

  • / Each
 

Description & Specifications

Specifications

Quantity 100g
Format Powder
Shelf Life 1825 Days
Additive Bile Salts No. 3
For Use With (Application) Microbiological culture media
Packaging Bottle
Certifications/Compliance ISO/DIS, 11014