Please login to your online account to display your discounted pricing

BD Difco™ Dehydrated Culture Media: Bile Salts No. 3

$569.49 - $1,110.03

Specifications

Additive Bile Salts No. 3
Certifications/Compliance ISO/DIS, 11014
Format Powder
Shelf Life 1825 Days
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

DF0130-17-4

 
bd
B213020
500g Each for $1,110.03

DF0130-15-6

 
bd
B213010
100g Each for $569.49
Description & Specifications

Specifications

Additive Bile Salts No. 3
Certifications/Compliance ISO/DIS, 11014
Format Powder
Shelf Life 1825 Days
Safety and Handling

1825 Days