Please login to your online account to display your discounted pricing

BD Difco™ Chromogenic Dehydrated Culture Media: Modified mTEC Agar


Culture media tests water for the presence of E.coli with membrane filtration method

Manufacturer: bd  B214884

 View more versions of this product

Catalog No. B14884

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2-25°C
Quantity 100g
Media Format No. 3 Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 10g/L, Sodium Chloride 7.5g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Lauryl Sulfate 0.2g/L, Sodium Desoxycholate 0.1g/L, 5-Bromo-6-chloro-3-indolyl-beta-D-glucuronide 0.5g/L, Agar 15g/L
Shelf Life 730 Days
Certifications/Compliance US, EPA
Color Light Beige
For Use With (Application) The chromogenic detection and enumeration of thermotolerant Escherichia coli in Water by the membrane filtration technique
Format Solid
Manufacturing Origin United States
Media Type Modified m TEC Agar
Packaging Bottle