Please login to your online account to display your discounted pricing

BD Difco™ Chromogenic Dehydrated Culture Media: MI Agar


Culture media tests drinking water for the presence of E.coli and total coliforms with membrane filtration method

Manufacturer: bd  B214882

 View more versions of this product

Catalog No. B14882

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2-25°C
Color Beige
For Use With (Application) To detect and enumerate Escherichia coli and total Coliforms in drinking Water by the membrane filtration technique
Format Solid
Manufacturing Origin United States
Media Format No. 3 Proteose Peptone 5g/L, Yeast extract 3g/L, D-Lactose 1g/L, 4-Methylumbelliferyl-beta-D-Galactopyranoside 0.1g/L, Indoxyl-beta-D-Glucuronide 0.32g/L, Sodium Chloride 7.5g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Lauryl Sulfate 0.2g/L, Sodium Desoxycholate 0.1g/L, Agar 15g/L
Media Type MI Agar
Packaging Bottle
Quantity 100g
Shelf Life 730 Days
Certifications/Compliance US, EPA