BD CultureSwab™ MaxV and MaxV+ Collection and Transport Systems
Improved recovery of fastidious organisms for proper patient diagnosis
Manufacturer:  BD 220233
 View more versions of this product
Catalog No. L220233
Description
Description
  • Improved recovery of fastidious organisms such as N. gonorrhoeae, H. influenzae, N. meningitidis, and S. pneumoniae
  • Sterile, ready-to-use systems intended for collection, transport, and preservation of clinical specimens for bacteriological examination
  • Medium is designed to maintain viability of organisms during transit to the laboratory
  • Pouches are made of a plastic film which retards penetration of atmospheric air into the product
Swabs contain unique blend of non-animal proteins embedded into swab fibers, to enhance recovery, in addition to all performance enhancing features of the BD CultureSwab line:
  • Nitrogen-flushed packaging
  • Patented tube design
  • Tamper-evident seal
  • Double-action security cap
  • Shaft: 5 1/4 in.
Specifications
Specifications
Stuart
Sterile
50 Pack
5 to 25°C
Calcium Chloride 0.1g/L, Mercaptoacetic Acid 1mL, Sodium Glycerophosphate 10g/L
Culture Swab™ MaxV Liquid Stuart
Nitrogen flushed
Single
Collection, transport and preservation of clinical specimens for Bacteriological examination
Documents