missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Sero-Fuge™ 2001 1-Speed Centrifuge

Choose a compact, digital centrifuge for your blood bank. The BD Sero-Fuge is Ideal for blood grouping, typing, cross matching, and other cell washing procedures.

Manufacturer:  BD B420351

 View more versions of this product

Catalog No. 05-100-1


 

Specifications

Description

Centrifuge; Sero-Fuge; Model 2001; 12-place rotor holds 12.7 x 75mm tubes; Fixed-speed for cell washing and serum separations; Speed range: 3100-3550rpm; Force: 1049xG; 115V

  • Keypad controls allow precise speed and run time settings
  • Four-digit display for run time (up to 99 minutes, 99 seconds) and time remaining
  • Stored parameters allow repeatable runs
  • On-board tachometer
  • Supplied rotor has numbered slots for easy sample identification
  • Variable-control brake
  • Locking lid prevents lid opening until rotor has reached a complete stop
  • Gasketed lid seal protects operator from exposure to aerosols within rotor chamber
  • Durable molded polycarbonate base and lid with a white polyurethane enamel finish
  • Four cushioned, rubber feet minimize sound and vibration
Description & Specifications

Specifications

12 75x13mm tubes
1999 x G
50/60Hz
Color Graphic Display
30.5cm
43cm
33cm
Keypad
12.7kg
115V 50/60Hz
3,100 to 3,550rpm
Variable-control brake
3550rpm
No
115V
12 in.
17 in.
13 in.
Fixed Angle
28 lb.
17 x 13 x 12 in. (43 x 33 x 30.5cm)
1,000 to 1,999 x G
No