Please login to your online account to display your discounted pricing

BD Bottled Media TAT Broth


For sterility and non-sterile product testing

Manufacturer: bd  B292250

 View more versions of this product

Catalog No. B4392255

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Fill Volume 190mL
For Use With (Application) Sterility and non-sterile product testing
Format Liquid
Manufacturing Origin United States
Packaging Bottle
Quantity 10 x 190mL