Please login to your online account to display your discounted pricing

BD Bottled Media Saboraud Dextrose Agar


For cultivation and maintenance of pathogenic and non-pathogenic fungi

Manufacturer: bd  B299510

 View more versions of this product

Catalog No. B4399510

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Quantity 500mL
Format Liquid; Without chloramphenicol
Manufacturing Origin United States
Packaging Bottle
Media Format Sabouraud Detrose Agar
Fill Volume 500mL
For Use With (Application) Used for qualitative procedures for cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes