missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD BBL™ Trypticase™ Soy Agar with Lecithin and Polysorbate 80 Sterile Pack Media - RODAC Plates

Plated media in gamma-irradiated packs for isolator systems—multiwrap packaging protects media from vaporized hydrogen peroxide exposure

$590.20 - $590.20

Specifications

Content And Storage 2°C to 8°C
Format Rodac
Media Type Rodac™, TSA With Lecithin and Polysorbate 80, Sterile Pack
Sterility Sterile
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/L, Polysorbate 80 5g/L, Agar 15g/L
View More Specs

Products 1
Catalog Number Mfr. No. Color Quantity Price Quantity  
Catalog Number Mfr. No. Color Quantity Price Quantity  
08-757-187
View Documents Promotion Details
BD
222207
Yellow 100/Pk.
Case of 100 Each for $590.20
 
Description

Description

Rodac (Replicate Organism Detection and Counting) plates for surface sampling.

To use:

  • Place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle
  • When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping
  • Use the included inner bag to transport plates from your critical environment

Compliance

Tested for performance and sterility according to Association for the Advancement of Medical Instrumentation (AAMI) guidelines.

Specifications

Specifications

2°C to 8°C
Rodac™, TSA With Lecithin and Polysorbate 80, Sterile Pack
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/L, Polysorbate 80 5g/L, Agar 15g/L
Gamma-irradiated Double Wrap
Rodac
Sterile
Monitoring surfaces in clean rooms and other environmentally controlled areas
SDS
missing translation for 'documents'

missing translation for 'documents'