Please login to your online account to display your discounted pricing

BD BBL™ Trypticase™ Dehydrated Culture Media Additive: Peptone


Manufacturer: bd  B211923

 View more versions of this product

Catalog No. B11923

  • / Each
 

Description & Specifications

Specifications

Packaging Pail
Format Powder
With Additives Peptone
Quantity 25 lb.
For Use With (Application) Dehydrated culture media
Certifications/Compliance LUO, MUO
Shelf Life 1460 Days
Accessories

1460 Days