Please login to your online account to display your discounted pricing

BD BBL™ Trypticase™ Dehydrated Culture Media Additive: Peptone


Manufacturer: bd  B211922

 View more versions of this product

Catalog No. B11922

  • / Each
 

Description & Specifications

Specifications

Packaging Pail
For Use With (Application) Dehydrated culture media
Format Powder
Quantity 5 lb.
Shelf Life 1460 Days
With Additives Peptone
Certifications/Compliance LUO, MUO
Quantity 5 lb.
Packaging Type Pail