Please login to your online account to display your discounted pricing

BD BBL™ Taxo™ P Discs (Cartridge)


For presumptive identification of pneumococci, and are intended for use with pure cultures

Manufacturer: bd  B231554

 View more versions of this product

 Disclaimers

For in vitro Diagnostic Use.

Catalog No. B31554

  • / Case of 10 Each
 

Description & Specifications

Specifications

Packaging 6 Cartridges
For Use With (Application) Differentiating Pneumococci
Concentration 5μg
Quantity 6 x 50 discs
Storage Requirements -20 to +8°C
Antimicrobial Agents Ethylhydrocupreine hydrochloride (optochin)
Shelf Life 1460 Days
Accessories

1460 Days