Please login to your online account to display your discounted pricing

BD BBL™ Taxo™ A Discs (Vial)


For presumptive identification of group A beta-hemolytic streptococci based on susceptibility to a low level of bacitracin

Manufacturer: bd  B231041

 View more versions of this product

 Disclaimers

For in vitro Diagnostic Use.

Catalog No. B31041

  • / Case of 6 Each
 

Description & Specifications

Specifications

Packaging 6 Vials
Storage Requirements -20 to +8°C
Concentration 0.04U
Shelf Life 548 Days
For Use With (Application) For differentiating Group A Streptococci
Quantity 6 x 50 discs
Antimicrobial Agents Bacitracin
Accessories

548 Days