Please login to your online account to display your discounted pricing

BD BBL™ Taxo™ A Discs (Vial)

For presumptive identification of group A beta-hemolytic streptococci based on susceptibility to a low level of bacitracin

$133.44 - $133.44

Specifications

Antimicrobial Agents Bacitracin
Concentration 0.04U
For Use With (Application) For differentiating Group A Streptococci
Shelf Life 548 Days
Storage Requirements -20 to +8°C
View More Specs

 Disclaimers

For in vitro Diagnostic Use.

Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B31041

 
bd
B231041
6 Vials 6 x 50 discs Case of 6 Each for $133.44
Description & Specifications

Specifications

Antimicrobial Agents Bacitracin
Concentration 0.04U
For Use With (Application) For differentiating Group A Streptococci
Shelf Life 548 Days
Storage Requirements -20 to +8°C

  • Impregnated with approximately 0.04 units of bacitracin per disc

Taxo A discs are not for susceptibility testing.

Vials of 50 discs

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.