BD BBL™ Taxo™ A Discs (Cartridge)
For presumptive identification of group A beta-hemolytic streptococci based on susceptibility to a low level of bacitracin
Supplier:  BD 231552
 View more versions of this product
Catalog No. B31552
Description
Description
  • Impregnated with approximately 0.04 units of bacitracin per disc
Taxo A discs are not for susceptibility testing.
Order Info
Cartridge of 50 discs
Specifications
Specifications
Bacitracin
-20°C to +8°C
10 Cartridges
0.04U
For differentiating Group A Streptococci
10 x 50 Discs
SDS
Documents
Documents
Promotions
Promotions
For in vitro Diagnostic Use.