Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Susceptibility Test Discs for Veterinary Laboratory Use


Same as standard Sensi-Disc Susceptibility Test Discs, but for veterinary laboratory use.

Manufacturer: bd  B291387

 View more versions of this product

Catalog No. B4391387

  • / Each
 

Description & Specifications

Specifications

Concentration 15μg
Antimicrobial Agents Tilmicosin
Packaging Disc
Quantity 50 Discs
Shelf Life 730 Days