Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Vancomycin (30mcg)


High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

Manufacturer: bd  B231353

 View more versions of this product

Catalog No. B31353

  • / Case of 10 Each
 

Description & Specifications

Specifications

Concentration 30μg
Antimicrobial Agents Vancomycin
Packaging Disc
Quantity 10 Cartridges
Shelf Life 1825 Days
Accessories

Accessories for Catalog Number: B31353

Item Mfr. No. Item Description Price Per Unit Quantity