Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Penicillin (10U)


High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

Manufacturer: bd  B231321

 View more versions of this product

Catalog No. B31321

  • / Case of 10 Each
 

Description & Specifications

Specifications

Antimicrobial Agents Penicillin
Concentration 10U
Packaging Disc
Quantity 10/Pk.
Shelf Life 730 Days
Accessories

Accessories for Catalog Number: B31321

Item Mfr. No. Item Description Price Per Unit Quantity  
14-890-8
BD BBL™ Sensi-Disc™ Antibiotic Susceptibility Test Discs Accessories: Sensi-Disc Dispenser
bd
B260661
Each for $1,977.74
14-890-6
BD BBL™ Sensi-Disc™ Antibiotic Susceptibility Test Discs Accessories: Sensi-Disc Dispenser
bd
B260660
Each for $1,914.00
B60640
BD BBL™ Sensi-Disc Designer Dispenser System
bd
B260640
Twelve-place self-tamping dispenser for Sensi-Disc discs Each for $2,704.59
14-890-5
BD BBL™ Sensi-Disc™ Antibiotic Susceptibility Test Disc Accessories: Single Disc Dispenser
bd
B260459
Case of 6 Each for $571.18
B60457
Becton Dickinson Cda Inc  SENSI-DISC DISPOBLE SINGLE
becton dickinson cda inc
B260457
Each for $87.92
Accessories

730 Days