Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Oxacillin

High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

$95.13 - $95.13

Specifications

Antimicrobial Agents Oxacillin
Concentration 1μg
Packaging Disc
Shelf Life 730 Days
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

B31319

  Accessories
bd
B231319
10/Pk. Case of 10 Each for $95.13
Description & Specifications

Specifications

Antimicrobial Agents Oxacillin
Concentration 1μg
Packaging Disc
Shelf Life 730 Days

  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.