Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Nalidixic Acid


High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

Manufacturer: bd  B231311

 View more versions of this product

Catalog No. B31311

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Antimicrobial Agents Nalidixic Acid
Packaging Disc
Quantity 10/Pk.
Shelf Life 1460 Days
Concentration 30μg
Accessories

Accessories for Catalog Number: B31311

Item Mfr. No. Item Description Price Per Unit Quantity