Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Minocyline


High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

Manufacturer: bd  B231251

 View more versions of this product

Catalog No. B31251

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Shelf Life 1095 Days
Concentration 30μg
Antimicrobial Agents Minocycline
Packaging Disc
Quantity 10/Pk.
Accessories

Accessories for Catalog Number: B31251

Item Mfr. No. Item Description Price Per Unit Quantity