Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Minocyline

High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

$22.72 - $96.28

Specifications

Antimicrobial Agents Minocycline
Concentration 30μg
Packaging Disc
Shelf Life 1095 Days
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

B31250

 
bd
B231250
Pack of 1 Cartridge; 50 Discs per Cartridge Each for $22.72

B31251

  Accessories
bd
B231251
10/Pk. Pack of 10 Each for $96.28
Description & Specifications

Specifications

Antimicrobial Agents Minocycline
Concentration 30μg
Packaging Disc
Shelf Life 1095 Days

  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration