Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Imipenem


High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

Manufacturer: bd  B231645

 View more versions of this product

Catalog No. B31645

  • / Case of 10 Each
 

Description & Specifications

Specifications

Concentration 10μg
Packaging Disc
Quantity 10/Pk.
Antimicrobial Agents Imipenem
Shelf Life 730 Days
Accessories

Accessories for Catalog Number: B31645

Item Mfr. No. Item Description Price Per Unit Quantity