Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Meropenem45


Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens

Manufacturer: bd  B231703

 View more versions of this product

 Disclaimers

For in vitro diagnostic use

Catalog No. B4331703

  • / Each
 

Description & Specifications

Specifications

Antimicrobial Agents Meropenem 45
Concentration 10μg
Packaging Disc
Quantity 1 Pack
Shelf Life 548 Days
Accessories

548 Days