Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Meropenem45


Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens

Manufacturer: bd  B231704

 View more versions of this product

 Disclaimers

For in vitro diagnostic use

Catalog No. B4331704

  • / Case of 10 Each
 

Description & Specifications

Specifications

Concentration 10μg
Antimicrobial Agents Meropenem 45
Packaging Disc
Quantity 10 Pack
Shelf Life 548 Days