Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cephalothin28


Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens

Manufacturer: bd  B231271

 View more versions of this product

 Disclaimers

For in vitro diagnostic use

Catalog No. B31271

  • / Case of 10 Each
 

Description & Specifications

Specifications

Packaging Disc
Concentration 30μg
Shelf Life 913 Days
Antimicrobial Agents Cephalothin28
Quantity 10 Pack
Accessories

Accessories for Catalog Number: B31271

Item Mfr. No. Item Description Price Per Unit Quantity