Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ceftriaxone26


Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens

Manufacturer: bd  B231635

 View more versions of this product

 Disclaimers

For in vitro diagnostic use

Catalog No. B31635

  • / Case of 10 Each
 

Description & Specifications

Specifications

Packaging Disc
Quantity 10 Pack
Antimicrobial Agents Ceftriaxone26
Concentration 30μg
Shelf Life 1095 Days
Accessories

Accessories for Catalog Number: B31635

Item Mfr. No. Item Description Price Per Unit Quantity  
14-890-8
BD BBL™ Sensi-Disc™ Antibiotic Susceptibility Test Discs Accessories: Sensi-Disc Dispenser
bd
B260661
Each for $1,977.74
14-890-6
BD BBL™ Sensi-Disc™ Antibiotic Susceptibility Test Discs Accessories: Sensi-Disc Dispenser
bd
B260660
Each for $1,914.00
B60640
BD BBL™ Sensi-Disc Designer Dispenser System
bd
B260640
Twelve-place self-tamping dispenser for Sensi-Disc discs Each for $2,704.59
14-890-5
BD BBL™ Sensi-Disc™ Antibiotic Susceptibility Test Disc Accessories: Single Disc Dispenser
bd
B260459
Case of 6 Each for $571.18
B60457
Becton Dickinson Cda Inc  SENSI-DISC DISPOBLE SINGLE
becton dickinson cda inc
B260457
Each for $87.92
Accessories

1095 Days