missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ceftriaxone26

Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens

$28.69 - $104.46

Specifications

Antimicrobial Agents Ceftriaxone26
Concentration 30μg
Packaging Disc
Shelf Life 1095 Days
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B31635 Accessories View Documents BD
B231635
10 Pk. Case of 10 Each for $104.46
 
B31634 View Documents BD
B231634
1 Pack Each for $28.69
 
Description

Description

  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content

Specifications

Specifications

Ceftriaxone26
30μg
Disc
1095 Days

For in vitro diagnostic use