Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ceftazidime/Clavulanic Acid


Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens

Manufacturer: bd  B231754

 View more versions of this product

 Disclaimers

For in vitro diagnostic use

Catalog No. B231754

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Packaging Disc
Shelf Life 365 Days
Concentration 10μg
Quantity 10 Pack
Antimicrobial Agents Ceftazidime, Clavulanic Acid
Accessories

365 Days