Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Aztreonam8


Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens

Manufacturer: bd  B231641

 View more versions of this product

 Disclaimers

For in vitro diagnostic use

Catalog No. B31641

  • / Case of 10 Each
 

Description & Specifications

Specifications

Quantity 10 Pack
Packaging Disc
Shelf Life 931 Days
Concentration 30μg
Antimicrobial Agents Aztreonam8
Accessories

931 Days