Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Disc, Ticarcillin w/Clavulanic Acid70


Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens

Manufacturer: bd  B231649

 View more versions of this product

 Disclaimers

For in vitro diagnostic use

Catalog No. B31649

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Concentration 75/10μg
Antimicrobial Agents Clavulanic Acid70
Packaging Disc
Quantity 10 Pack
Shelf Life 548 Days