Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Antimicrobial Discs


Allow addition of antimicrobial agents to culture media.

Manufacturer: bd  B231574

 View more versions of this product

 Disclaimers

Not for use in ordinary susceptibility plate tests.

Catalog No. B31574

  • / Each
 

Description & Specifications

Specifications

Concentration 50μg
Quantity Each
Antimicrobial Agents p-Aminosalicylic Acid
Packaging Disc
Shelf Life 1095 Days