Please login to your online account to display your discounted pricing

BD BBL™ Sensi-Disc™ Antimicrobial Discs


Allow addition of antimicrobial agents to culture media.

Manufacturer: bd  B231573

 View more versions of this product

 Disclaimers

Not for use in ordinary susceptibility plate tests.

Catalog No. B31573

  • / Each
 

Description & Specifications

Specifications

Quantity Each
Packaging Disc
Antimicrobial Agents p-Aminosalicylic Acid
Shelf Life 1095 Days
Concentration 10μg
Accessories

1095 Days