Please login to your online account to display your discounted pricing

BD BBL™ Replacement Reagents for Gram Stain Kits


For BBL Gram Stain Kits

Manufacturer: bd  B212532

 View more versions of this product

Catalog No. B4312532

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 15° to 30°C
Stain Type Safranin
Type Gram Safranin
Quantity 3.8L
Certifications/Compliance CE
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method