Please login to your online account to display your discounted pricing

BD BBL™ Replacement Reagents for Gram Stain Kits


For BBL Gram Stain Kits

Manufacturer: bd  B212544

 View more versions of this product

Catalog No. B4312544

  • / Pack of 4 Each
 

Description & Specifications

Specifications

Storage Requirements 15° to 30°C
Stain Type Basic Fuchsin
Type Gram Basic Fuchsin
Quantity 4 x 250mL
Certifications/Compliance CE
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method