missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD BBL™ Prepared Sterile Pack Settling Plated Trypticase™ Soy Agar

Plated media in gamma-irradiated packs for isolator systems-multiwrap packaging protects media from vaporized hydrogen peroxide exposure

Manufacturer:  BD 222205

 View more versions of this product

Catalog No. 08-757-188



Description

Description

  • Available in 100 x 15 and 150 x 15mm style plates

Specifications

Specifications

ISO/DIS, 11014
2°C to 8°C
US
Sterile
100/Pk.
Light Beige
100 x 15mm Settling
Trypticase™ Soy Agar
Plate
SDS
Documents

Documents

Product Certifications
Promotions

Promotions