Please login to your online account to display your discounted pricing

BD BBL™ Prepared Sterile Pack Settling Plated Trypticase™ Soy Agar


Plated media in gamma-irradiated packs for isolator systems-multiwrap packaging protects media from vaporized hydrogen peroxide exposure

Manufacturer: bd  B222206

 View more versions of this product

Catalog No. 08-757-189

  • / Case of 45 Each
 

Description & Specifications

Specifications

Quantity 45/Pk.
Format 150 x 15mm Settling
Storage Requirements 2-8°C
Sterile Yes
Packaging Plate
Certifications/Compliance ISO/DIS, 11014
Manufacturing Origin United States
Color Light Beige
Media Type Trypticase™ Soy Agar
Shelf Life 154 Days
Accessories

154 Days

SDS & Safety Documents