Please login to your online account to display your discounted pricing

BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (TSA II™) with Sheep Blood


Used for cultivating fastidious microorganisms and for visualization of hemolytic reactions produced by many bacterial species

Manufacturer: bd  B221239

 View more versions of this product

Catalog No. B21239

  • / Pack of 20 Each
 

Description & Specifications

Specifications

Storage Requirements 2-8°C
Packaging Plate
Certifications/Compliance ISO/DIS, 11014
Color Red
For Use With (Application) Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Manufacturing Origin United States
Media Format Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Defibrinated Sheep Blood 5% or 10%
Media Type Trypticase™ Soy Agar with 5% Sheep Blood (TSA II)
Quantity 20/Pk.
Shelf Life 105 Days