Please login to your online account to display your discounted pricing

BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (Soybean-Casein Digest Agar)


Used for isolation and cultivation of non-fastidious and fastidious microorganisms

Manufacturer: bd  B221185

 View more versions of this product

Catalog No. B21185

  • / Pack of 20 Each
 

Description & Specifications

Specifications

Media Format Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L
Color Light Beige
Format Liquid
Manufacturing Origin United States
Packaging Plate
Quantity 20/Pk.
Shelf Life 112 Days
Storage Requirements 2-8°C
Certifications/Compliance ISO/DIS, 11014
For Use With (Application) Cultivation of Non-fastidious and fastidious microorganisms (not for anaerobes)
Media Type Trypticase™ Soy Agar (Soybean Casein digest Agar)