BD BBL™ Prepared Plated Media: Thayer Martin, Modified (MTM II) Agar
For the isolation of pathogenic Neisseria from specimens containing mixed flora of bacteria and fungi
Manufacturer:  BD B221567
 View more versions of this product
Catalog No. B21567X
Description
Description
  • Ready for immediate use
Specifications
Specifications
Dark Brown
112 Days
Liquid
ISO/DIS, 11014
United States
20/Pk.
Thayer Martin, modified (MTM II) Agar
Plate