Please login to your online account to display your discounted pricing

BD BBL™ Prepared Plated Media: Sabouraud Dextrose Agar (Deep Fill)


Used in qualitative procedures for cultivation of dermatophytes

Manufacturer: bd  B221180

 View more versions of this product

Catalog No. B21180

  • / Pack of 20 Each
 

Description & Specifications

Specifications

Media Format Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L
Packaging Plate
Manufacturing Origin United States
Color Light Beige
Format Solid
Quantity 20/Pk.
Certifications/Compliance USP, ISO/DIS, 11014
Media Type Sabouraud Dextrose Agar (deep fill)
For Use With (Application) Qualitative procedures for cultivation of Dermatophytes, Candida albicans
Shelf Life 147 Days
Accessories

147 Days