missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ Prepared Plated Media: Martin-Lewis Agar
For the isolation of pathogenic Neisseria from specimens containing mixed flora of bacteria and fungi
$44.60 - $44.60
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Color | Dark Brown |
Format | Liquid |
For Use With (Application) | Pathogenic Neisseria |
Media Type | Martin-Lewis Agar |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B21557X
|
BD
B221557 |
20/Pk. |
Pack of 20 Each for $44.60
|
|
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
ISO/DIS, 11014 | |
Liquid | |
Martin-Lewis Agar | |
United States | |
112 Days |
Dark Brown | |
Pathogenic Neisseria | |
Plate | |
Pancreatic digest of Casein 7.5g/L, Selected meat Peptone 7.5g/L, Corn Starch 1g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 5g/L, Agar 12g/L, Hemoglobin 10g/L, IsoVitaleX™ enrichment 10mL, V-C-A Inhibito |