Please login to your online account to display your discounted pricing

BD BBL™ Prepared Plated Media: Martin-Lewis Agar


For the isolation of pathogenic Neisseria from specimens containing mixed flora of bacteria and fungi

Manufacturer: bd  B221557

 View more versions of this product

Catalog No. B21557X

  • / Pack of 20 Each
 

Description & Specifications

Specifications

Quantity 20/Pk.
Color Dark Brown
For Use With (Application) Pathogenic Neisseria
Format Liquid
Manufacturing Origin United States
Media Format Pancreatic digest of Casein 7.5g/L, Selected meat Peptone 7.5g/L, Corn Starch 1g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 5g/L, Agar 12g/L, Hemoglobin 10g/L, IsoVitaleX™ enrichment 10mL, V-C-A Inhibito
Media Type Martin-Lewis Agar
Packaging Plate
Shelf Life 112 Days
Weight (Metric) 1200g
Certifications/Compliance ISO/DIS, 11014