Please login to your online account to display your discounted pricing

BD BBL™ Prepared Plated Media: Columbia CNA Agar with 5% Sheep Blood


Selective and differential media used for the isolation and differentiation of gram-positive microorganisms from clinical and nonclinical materials

Manufacturer: bd  B221352

 View more versions of this product

Catalog No. B221352

  • / Pack of 20 Each
 

Description & Specifications

Specifications

Quantity 20/Pk.
Storage Requirements 2-8°C
Weight (Metric) 1300g
Color Red
Format Liquid
Packaging Plate
Certifications/Compliance ISO/DIS, 11014
Manufacturing Origin United States
Media Type Columbia Colistin-Naladixic Acid Agar with 5% Sheep Blood
For Use With (Application) Gram positive microorganisms
Media Format Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3.5g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Colistin 10mg/L, Nalidixic Acid 10mg/L, Sheep Blood, defibrinated 5%
Shelf Life 84 Days
Accessories

84 Days