Please login to your online account to display your discounted pricing

BD BBL™ Prepared Plated Media: Brain Heart Infusion Agar (Deep Fill)


For cultivation of a wide variety of organism types, including bacteria, yeasts and molds

Manufacturer: bd  B221569

 View more versions of this product

Catalog No. B21569X

  • / Pack of 20 Each
 

Description & Specifications

Specifications

Quantity 20/Pk.
Storage Requirements 2-25°C
Color Light Beige
For Use With (Application) Cultivation of wide variety of organism types, including bacteria, Yeasts and molds
Format Liquid
Manufacturing Origin United States
Media Format Brain heart infusion from (solids) 8g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 16g/L, Sodium Chloride 5g/L, Dextrose 2g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L
Media Type Brain heart infusion Agar (deep fill)
Packaging Plate
Shelf Life 112 Days
Certifications/Compliance ISO/DIS, 11014