BD BBL™ Prepared Plated Media: Brain Heart Infusion Agar (Deep Fill)
For cultivation of a wide variety of organism types, including bacteria, yeasts and molds
$53.67 - $53.67
Specifications
Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Format Liquid
Media Type Brain heart infusion Agar (deep fill)
Packaging Plate
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Application) Quantity Storage Requirements Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Quantity Storage Requirements Price Quantity  
B21569X
View Documents
BD
221569
Cultivation of wide variety of organism types, including bacteria, Yeasts and molds 20/Pk. 2-25°C
Pack of 20 Each for $53.67
Description
Description
  • To reduce the effects of drying during prolonged incubation, the plates can be deep filled
Specifications
Specifications
ISO/DIS, 11014
Liquid
Plate
Brain heart infusion from (solids) 8g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 16g/L, Sodium Chloride 5g/L, Dextrose 2g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L
Light Beige
Brain heart infusion Agar (deep fill)
United States
112 Days
SDS