missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ Prepared Plated Media: Bordet Gengou Blood Agar
For detection and isolation of Bordetella pertussis from clinical specimens
Supplier: BD 297876
Specifications
ISO/DIS, 11014 | |
Liquid | |
Bordet Gengou Blood Agar | |
Bordetella pertussis from clinical specimens | |
10/Pk. |
Red | |
United States | |
Potato infusion from (solids) 4.5g/L, Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Sodium Chloride 5.5g/L, Agar 20g/L, Glycerol 10mL, Sheep Blood, defibrinated 15% | |
Plate | |
2800 g |